Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYROXD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584757 Supplier Invitrogen™ Supplier No. PA584757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLTAPITKVL DQWFESEPLK ATLATDAVIG AMTSPHTPGS GYVLLHHVMG GLEGMQGAWG YVQGGMGALS DAIASSATTH GASIFTE Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 95%.

PYROXD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYROXD2
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PYROXD2
Gene Accession No. Q8N2H3
Gene Alias 3830409H07Rik; 4833409A17Rik; C10orf33; FP3420; Phytn_dehydro and Pyr_redox domain containing protein RGD1303232; pyridine nucleotide-disulfide oxidoreductase domain-containing protein 2; pyridine nucleotide-disulphide oxidoreductase domain 2; Pyroxd2; RGD1303232
Gene Symbols PYROXD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PYROXD2. Recombinant protein control fragment (Product #RP-105424).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84795
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.