Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PYROXD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113942
Description
Immunogen sequence: MAASGRGLCK AVAASPFPAW RRDNTEARGG LKPEYDAVVI GAGHNGLVAA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
PYROXD2 is a protein coding gene.
Specifications
PYROXD2 | |
Polyclonal | |
Unconjugated | |
PYROXD2 | |
3830409H07Rik; 4833409A17Rik; C10orf33; FP3420; Phytn_dehydro and Pyr_redox domain containing protein RGD1303232; pyridine nucleotide-disulfide oxidoreductase domain-containing protein 2; pyridine nucleotide-disulphide oxidoreductase domain 2; Pyroxd2; RGD1303232 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84795 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N2H3 | |
PYROXD2 | |
Synthetic peptide directed towards the N terminal region of human C10orf33 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction