Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Pyruvate Carboxylase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578100
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Pig (97%), Chicken (84%), Rhesus Monkey (100%), Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pyruvate Carboxylase requires biotin and ATP to catalyse the carboxylation of pyruvate to oxaloacetate. The active enzyme is a homotetramer arranged in a tetrahedron which is located exclusively in the mitochondrial matrix. Pyruvate carboxylase is involved in gluconeogenesis, lipogenesis, insulin secretion and synthesis of the neurotransmitter glutamate. Mutations in this gene have been associated with pyruvate carboxylase deficiency.
Specifications
Pyruvate Carboxylase | |
Polyclonal | |
Unconjugated | |
PC | |
EC 6.4.1.1; Pc; Pcb; Pcx; PYC; pyruvate carboxylase; pyruvate carboxylase, mitochondrial; pyruvate decarboxylase; pyruvic carboxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25104, 5091 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.69 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P11498, P52873 | |
PC | |
Recombinant protein encompassing a sequence within the C-terminus region of human Pyruvate Carboxylase. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction