Learn More
Invitrogen™ Pyruvate Carboxylase Polyclonal Antibody
Description
Keep as concentrated solution. Predicted reactivity: Rat (100%), Pig (100%), Rhesus Monkey (100%), Bovine (100%). Positive Control: 293T, A431, HeLa, mouse liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Pyruvate Carboxylase |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3.08 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | PC |
| Gene Accession No. | P11498, Q05920 |
| Gene Alias | EC 6.4.1.1; Pc; Pcb; Pcx; PYC; pyruvate carboxylase; pyruvate carboxylase, mitochondrial; pyruvate decarboxylase; pyruvic carboxylase |
| Gene Symbols | PC, Pcx |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.