Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pyruvate Carboxylase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566203
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566203 Supplier Invitrogen™ Supplier No. PA566203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAHFKDFTATF GPLDSLNTRL FLQGPKIAEE FEVELERGKT LHIKALAVSD LNRAGQRQVF FELNGQLRSI LVKDTQAMKE MHFHPKALKD VKGQIG Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

Pyruvate Carboxylase requires biotin and ATP to catalyse the carboxylation of pyruvate to oxaloacetate. The active enzyme is a homotetramer arranged in a tetrahedron which is located exclusively in the mitochondrial matrix. Pyruvate carboxylase is involved in gluconeogenesis, lipogenesis, insulin secretion and synthesis of the neurotransmitter glutamate. Mutations in this gene have been associated with pyruvate carboxylase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pyruvate Carboxylase
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PC
Gene Accession No. P11498
Gene Alias EC 6.4.1.1; Pc; Pcb; Pcx; PYC; pyruvate carboxylase; pyruvate carboxylase, mitochondrial; pyruvate decarboxylase; pyruvic carboxylase
Gene Symbols PC
Host Species Rabbit
Immunogen Recombinant Human Pyruvate Carboxylase. Recombinant protein control fragment (Product #RP-105110).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.