Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Carboxylase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575545 Supplier Thermo Scientific Supplier No. PA575545
Add to Cart
Add to Cart

Pyruvate Carboxylase Polyclonal Antibody

Pyruvate Carboxylase Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Pyruvate Carboxylase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PC
Gene Accession No. P11498, P52873, Q05920
Gene Alias EC 6.4.1.1; Pc; Pcb; Pcx; PYC; pyruvate carboxylase; pyruvate carboxylase, mitochondrial; pyruvate decarboxylase; pyruvic carboxylase
Gene Symbols PC, Pcx
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 400-445 of Human Pyruvate Carboxylase.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18517, 18563, 25104, 5091
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.