Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Pyruvate Carboxylase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572953
Description
Pyruvate Carboxylase Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Pyruvate Carboxylase requires biotin and ATP to catalyse the carboxylation of pyruvate to oxaloacetate. The active enzyme is a homotetramer arranged in a tetrahedron which is located exclusively in the mitochondrial matrix. Pyruvate carboxylase is involved in gluconeogenesis, lipogenesis, insulin secretion and synthesis of the neurotransmitter glutamate. Mutations in this gene have been associated with pyruvate carboxylase deficiency.
Specifications
Pyruvate Carboxylase | |
Polyclonal | |
Unconjugated | |
PC | |
EC 6.4.1.1; Pc; Pcb; Pcx; PYC; pyruvate carboxylase; pyruvate carboxylase, mitochondrial; pyruvate decarboxylase; pyruvic carboxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18517, 18563, 25104, 5091 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.0 mg/mL | |
PBS with 0.02% sodium azide | |
P11498, P52873, Q05920 | |
PC, Pcx | |
Partial recombinant protein made to an internal region of the human Pyruvate Carboxylase protein (within residues 930-1050). [Swiss-Prot P11498]. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction