Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvate Dehydrogenase E1-alpha subunit Antibody (006), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP306600AF488
Description
Pyruvate Dehydrogenase E1-alpha subunit Monoclonal antibody specifically detects Pyruvate Dehydrogenase E1-alpha subunit in Human samples. It is validated for ImmunofluorescenceSpecifications
Pyruvate Dehydrogenase E1-alpha subunit | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
006 | |
Immunocytochemistry/Immunofluorescence | |
EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Pyruvate Dehydrogenase E1-alpha subunit (P08559-1; Phe30-Ser390). | |
0.1 mL | |
Cancer, Core ESC Like Genes, Lipid and Metabolism, Phospho Specific, Signal Transduction, Stem Cell Markers | |
5160 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction