Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvate Dehydrogenase E1-alpha subunit Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25474725UL
Description
Pyruvate Dehydrogenase E1-alpha subunit Polyclonal specifically detects Pyruvate Dehydrogenase E1-alpha subunit in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Pyruvate Dehydrogenase E1-alpha subunit | |
Polyclonal | |
Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
PDHA1 | |
This antibody was developed against a Recombinant Protein corresponding to amino acids:YCRSGKGPILMELQTYRYHGHSMSDPGVS | |
25 μL | |
Cancer, Core ESC Like Genes, Lipid and Metabolism, Signal Transduction, Stem Cell Markers | |
5160 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction