Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvate Dehydrogenase Kinase 1/PDK1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002384AF700
Description
Pyruvate Dehydrogenase Kinase 1/PDK1 Polyclonal specifically detects Pyruvate Dehydrogenase Kinase 1/PDK1 in Human, Mouse samples. It is validated for Western Blot, Immunoblotting.Specifications
Pyruvate Dehydrogenase Kinase 1/PDK1 | |
Polyclonal | |
Western Blot, Immunoblotting | |
Q15118 | |
PDK1 | |
A synthetic peptide made to an internal portion of the human PDK1 protein sequence (between residues 300-400). [Swiss-Prot Q15118] | |
Affinity Purified | |
RUO | |
Primary | |
Human. Predicted to react with rat, mouse, primate and bovine based on 100% sequence homology. Immunogen sequence has 92% homology to chicken, 91% homology to Zebrafish and porcine. | |
Store at 4C in the dark. |
Western Blot, Immunoblot | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial | |
Rabbit | |
49 kDa | |
0.1 mL | |
Autophagy, Cancer, Cytoskeleton Markers, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
5163 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction