Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvate Dehydrogenase Kinase 1/PDK1 Rabbit anti-Human, Mouse, Rat, Clone: 9A7X4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Pyruvate Dehydrogenase Kinase 1/PDK1 |
---|---|
Clone | 9A7X4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Pyruvate Dehydrogenase Kinase 1/PDK1 Monoclonal antibody specifically detects Pyruvate Dehydrogenase Kinase 1/PDK1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Pyruvate Dehydrogenase Kinase 1/PDK1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5163 | |
IgG | |
Affinity purified |
9A7X4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial | |
A synthesized peptide derived from human Pyruvate Dehydrogenase Kinase 1/PDK1/PDHK1 | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title