Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Kinase Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990451
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0451 100 μg
50-199-0452 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0451 Supplier Rockland Immunochemicals Supplier No. 20011780100
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-PYRUVATE KINASE is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum as well as purified and partially purified Pyruvate Kinase (Rabbit Muscle). This product has been reported to react with all forms of pyruvate kinase (pan M-PK). Cross-reactivity against pyruvate kinase from other mammalian tissues is expected but has not been specifically determined.

PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.

Specifications

Antigen Pyruvate Kinase
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene PKM
Gene Accession No. P11974
Gene Alias AA414905; AL024370; AL024424; CTHBP; Cytosolic thyroid hormone-binding protein; epididymis secretory protein Li 30; HEL-S-30; isozymes M1/M2; OIP3; OIP-3; Opa-interacting protein 3; p58; PK, muscle type; PK2; Pk-2; PK3; Pk-3; PKM; pkm.L; PKM12; PKM2; Pykm; Pyruvate kinase 2/3; pyruvate kinase 3; pyruvate kinase isozymes M1/M2; pyruvate kinase M1/2 L homeolog; pyruvate kinase M1/2 L homeolog; pyruvate kinase, muscle; Pyruvate kinase muscle isozyme; Pyruvate kinase PKM; pyruvate kinase, muscle; TCB; THBP1; Thyroid hormone-binding protein 1; thyroid hormone-binding protein, cytosolic; tu; Tumor M2-PK; XELAEV_18018020mg
Gene Symbols PKM
Host Species Goat
Immunogen Pyruvate Kinase (Rabbit Muscle)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008676
Target Species Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.