Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Glass bottle
127-17-3
C3H4O3
88.06
MFCD00002585
LCTONWCANYUPML-UHFFFAOYSA-N
pyruvic acid,pyroracemic acid,2-oxopropionic acid,acetylformic acid,alpha-ketopropionic acid,2-ketopropionic acid,2-oxopropanoate,propanoic acid, 2-oxo,2-oxo-propionic acid,2-oxopropansaeure
1060
CHEBI:32816
2-oxopropanoic acid
CC(=O)C(=O)O
127-17-3
C3H4O3
88.06
MFCD00002585
LCTONWCANYUPML-UHFFFAOYSA-N
pyruvic acid,pyroracemic acid,2-oxopropionic acid,acetylformic acid,alpha-ketopropionic acid,2-ketopropionic acid,2-oxopropanoate,propanoic acid, 2-oxo,2-oxo-propionic acid,2-oxopropansaeure
1060
CHEBI:32816
2-oxopropanoic acid
CC(=O)C(=O)O
Melting Point 11.8°C
Specific Gravity 1.25
Packaging Glass bottle
Refractive Index 1.4260 to 1.4300
Quantity 25g
Flash Point 82°C
Infrared Spectrum Authentic
Color Amber or Colorless to Yellow
Boiling Point 165.0°C
Density 1.2500g/mL
Assay Percent Range 98%
Linear Formula CH3COCO2H
Beilstein 03, 608
Fieser 01,974
Merck Index 15, 8135
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether
Formula Weight 88.06
Physical Form Liquid
Percent Purity 98%
Grade Extra Pure
Chemical Name or Material Pyruvic acid, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

EINECSNumber : 204-824-3

RTECSNumber : UZ0829800

TSCA : TSCA