Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvic acid, 98%, extra pure

Catalog No. AC132145000
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 127-17-3 | C3H4O3 | 88.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-17-3
Molecular Formula C3H4O3
Molecular Weight (g/mol) 88.06
MDL Number MFCD00002585
InChI Key LCTONWCANYUPML-UHFFFAOYSA-N
Synonym pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure
PubChem CID 1060
ChEBI CHEBI:32816
IUPAC Name 2-oxopropanoic acid
SMILES CC(=O)C(O)=O

Specifications

Melting Point 11.8°C
Color Amber or Colorless to Yellow
Density 1.2500g/mL
Boiling Point 165.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4260 to 1.4300
Linear Formula CH3COCO2H
Quantity 500 g
Beilstein 03, 608
Fieser 01,974
Specific Gravity 1.25
Merck Index 15, 8135
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and ether
Formula Weight 88.06
Percent Purity 98%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Pyruvic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 204-824-3
RTECSNumber UZ0829800
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.