Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvic acid sodium salt, 99.3%, Cell Culture Reagent, MP Biomedicals™

Catalog No. icn19473425 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. ICN19473425

Supplier: MP Biomedicals, Inc 0219473425

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.044
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 110
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material Pyruvic acid sodium salt, Molecular Biology
EINECSNumber 204-024-4
Recommended Storage Store at +4°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.