Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvic aldehyde, 30-45 wt% solution in water
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -20.0°C |
| Color | Brown-Yellow to Yellow |
| Density | 1.2000g/mL |
| pH | 1 to 3 |
| Boiling Point | >72.0°C |
| Flash Point | >71°C |
| Packaging | Glass bottle |
| Refractive Index | 1.4209 |
| Linear Formula | CH3COCHO |
| Quantity | 100 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Suspected of causing genetic defects. May cause an allergic skin reaction. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Con |
| EINECSNumber | 201-164-8 |
| RTECSNumber | UZ0700000 |
| TSCA | TSCA |
| Recommended Storage | 4°C |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction