Learn More
Invitrogen™ PYY Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595608
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - IHC: human appendicitis tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Peptide tyrosine-tyrosine amide (3-36), or PYY (3-36), is a major metabolite of the gut hormone PYY and is produced by the action of dipeptidyl peptidase IV on PYY in the intestinal brush border and in the circulation. PYY (3-36) is a potent inhibitor of food intake in rats and humans, acting selectively on the Y2 receptor to inhibit orexigenic NPY neurons in the arcuate nucleus, thus disinhibiting anorexigenic POMC neurons. The anorexigenic effect of PYY (3-36) is additive to that of the co-secreted gut hormone GLP-1.
Specifications
PYY | |
Polyclonal | |
Unconjugated | |
Pyy | |
GHYY; Peptide tyrosine tyrosine; peptide tyrosine-tyrosine (YY); peptide YY; peptide YY (mapped); Peptide YY(3-36); Peptide YY3-36; Peptide YY3-37; peptide-YY; prepro-PYY; PYY; PYY II; PYY1; PYY-1; PYY-2; PYYI; PYY-I; PYYII; PYY-II; RATGHYY; Yy | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5697 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
P10082 | |
Pyy | |
A synthetic peptide corresponding to a sequence of human Peptide YY/PYY (YPIKPEAPREDASPEELNRYYASLRHYLNLVTRQRY). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.