Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PYY Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. 01674464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-464 Supplier Invitrogen™ Supplier No. PA19590
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PA1-9590 detects PYY in human, mouse and rat samples. PA1-9590 has been successfully used in ELISA procedures. PA1-9590 immunogen is a synthetic peptide corresponding to amino acids 3-18 (I K P E A P G E D A S P E E L N) of human PYY.

Peptide tyrosine-tyrosine amide (3-36), or PYY (3-36), is a major metabolite of the gut hormone PYY and is produced by the action of dipeptidyl peptidase IV on PYY in the intestinal brush border and in the circulation. PYY (3-36) is a potent inhibitor of food intake in rats and humans, acting selectively on the Y2 receptor to inhibit orexigenic NPY neurons in the arcuate nucleus, thus disinhibiting anorexigenic POMC neurons. The anorexigenic effect of PYY (3-36) is additive to that of the co-secreted gut hormone GLP-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PYY
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.02% sodium azide
Gene Pyy
Gene Accession No. P10082, P10631, Q9EPS2
Gene Alias GHYY; Peptide tyrosine tyrosine; peptide tyrosine-tyrosine (YY); peptide YY; peptide YY (mapped); Peptide YY(3-36); Peptide YY3-36; Peptide YY3-37; peptide-YY; prepro-PYY; PYY; PYY II; PYY1; PYY-1; PYY-2; PYYI; PYY-I; PYYII; PYY-II; RATGHYY; Yy
Gene Symbols Pyy
Host Species Chicken
Immunogen Synthetic peptide corresponding to amino acids 3-18 of human PYY.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217212, 287730, 5697
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.