Learn More
Invitrogen™ PZP Polyclonal Antibody
Description
PZP Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PZP |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | PZP |
| Gene Accession No. | Q63041 |
| Gene Alias | A1m; A1m {ECO:0000312; A2m; AI893533; alpha 1 macroglobulin; alpha-1-M; Alpha-1-macroglobulin; alpha-1-macroglobulin 165 kDa subunit; Alpha-1-macroglobulin 45 kDa subunit; alpha-2-M; alpha-2-macroglobulin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6; CPAMD6; EMBL:AAA40723.1}; MAM; Pregnancy zone protein; pregnancy-zone protein; Pzp; PZP, alpha-2-macroglobulin like |
| Gene Symbols | PZP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.