Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PZP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559554 Supplier Invitrogen™ Supplier No. PA559554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFSINTTSIS VNKLFVRVFT VHPNLCFHYS WVAEDHQGAQ HTANRVFSLS GSYIHLEPVA GTLPCGHTET ITAHYT Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 46%.

The protein encoded by this gene is highly expressed in late-pregnancy serum and is similar in structure to alpha-2-macroglobulin. The encoded protein, which acts as a homotetramer, inhibits the activity of all four classes of proteinases. This protein contains cleavage sites for several proteinases. Upon binding of a proteinase, the conformation of this protein changes to trap the proteinase, limiting its activity. This protein appears to be elevated in the sera of presymptomatic Alzheimer's disease patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PZP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PZP
Gene Accession No. P20742
Gene Alias A1m; A1m {ECO:0000312; A2m; AI893533; alpha 1 macroglobulin; alpha-1-M; Alpha-1-macroglobulin; alpha-1-macroglobulin 165 kDa subunit; Alpha-1-macroglobulin 45 kDa subunit; alpha-2-M; alpha-2-macroglobulin; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6; CPAMD6; EMBL:AAA40723.1}; MAM; Pregnancy zone protein; pregnancy-zone protein; Pzp; PZP, alpha-2-macroglobulin like
Gene Symbols PZP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PZP. Recombinant protein control fragment (Product #RP-98419).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5858
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.