Learn More
OraSure Technologies Q.E.D.™ Saliva Alcohol Test, A-150
SDP
Catalog No. 23222109 Shop All OraSure Technologies Products
Click to view available options
Quantity:
100 Tests
30 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy
The Q.E.D.™ Saliva Alcohol Test is the 'on-the-spot leader' in the rapid, accurate and quantitative determination of alcohol in saliva. With a high correlation to blood-alcohol levels, Q.E.D. has a testing range of 0-145 mg/dL and provides quantitative results in minutes.
  • Reliable, Fast, Simple
  • Non-invasive
  • Quantitative results in minutes
  • Reads like a thermometer
  • No instrumentation
  • Long shelf life
  • Built-in QA Spot™ assures quality control for every test
Specifications
Certifications/Compliance CLIA-waived and DOT-approved alcohol testing
Type Saliva Alcohol Test
For Use With (Application) On-Site, Low-Cost Alternative to Breath or Blood Alcohol Testing
Quantity 100 Tests
Test Time <5 min.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.