Learn More
Restek™ Q-Sep QuEChERS Extraction Salts
SDP
Catalog No. 06611921 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
4 g MgSO4, 1 g NaCl
4 g MgSO4, 1 g NaCl, 1 g TSCD, 0.5 g DHS
6 g MgSO4, 1.5 g NaOAc
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-611-921 4 g MgSO4, 1 g NaCl
06-611-913 6 g MgSO4, 1.5 g NaOAc
06-611-949 4 g MgSO4, 1 g NaCl, 1 g TSCD, 0.5 g DHS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-611-921 Supplier Restek™ Supplier No. 25847
Add to Cart
Add to Cart
Pre-weighed, ultra-pure extraction salts
  • Easy-open packets eliminate need for a second empty tube for salt transfer
  • Free-flowing salts transfer easily and completely
  • Convenient slim packets fit perfectly into tubes to prevent spills
  • Ready-to-use tubes, no glassware required
  • Ideal for original unbuffered, AOAC (2007.01), and European (EN 15662) QuEChERS methods
Specifications
Type Q-Sep QuEChERS Extraction Salts
Includes Salt packets
Material 4 g MgSO4, 1 g NaCl
Quantity 50 packets