Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Q-VD-OPH Inhibitor Peptide
SDP

Catalog No. NBP229391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229391 1 mg
NBP2293913 3 mg
NBP2293915 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP229391 Supplier Novus Biologicals™ Supplier No. NBP229391
Add to Cart
Add to Cart

Applications: Functional, In vitro assay

Specifications

Host Species Human
Color Off-white
For Use With (Application) Functional, In vitro assay
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 1 mg
Product Type Q-VD-OPH Inhibitor Peptide
Molecular Weight (g/mol) 513
Inhibitors Q-VD-OPH
Form Semi-solid

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.