Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qa-1(b) Mouse, anti-Mouse, R718, Clone: 6A8.6F10.1A6, BD OptiBuild™

Catalog No. BDB752126 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752126 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752126 Supplier BD Biosciences Supplier No. 752126
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 6A8.6F10.1A6 monoclonal antibody reacts with the Qa-1[b] alloantigen, which is a nonclassical MHC class I (Class Ib) molecule encoded by the T23 gene of the H-2 complex (H2-T23). Qa-1 associates with β2-microglobulin and is expressed at low levels on most leukocytes and many other cell types. Its level of cell-surface expression is upregulated by IFNγ or specific peptides. Qa-1 is an oligomorphic molecule which presents a limited pool of peptides to T lymphocytes bearing αβ and γδTCR and binds to a large subpopulation of NK cells. In fact, Qa-1[b] is the ligand for CD94/NKG2A, CD94/NKG2C, and CD94/NKG2E receptors, which are expressed on NK cells. Furthermore, it has been reported that Qa-1 expressed on activated B lymphocytes is involved in immunoregulation by inducing T-cell-mediated suppression of antibody responses. The 6A8.6F10.1A6 mAb can detect Qa-1[b] on activated splenocytes from C57BL/6 and BALB/c mice (both Qa-1[b]), but not from A/j mice (Qa-1[a]), and it can block target cell recognition by CTLs specific for Qa-1-presented antigen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Qa-1(b)
Applications Flow Cytometry
Classification Monoclonal
Clone 6A8.6F10.1A6
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.