Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Qa-2 Monoclonal Antibody (695H1-9-9)
GREENER_CHOICE

Catalog No. PIMA528662
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528662 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528662 Supplier Invitrogen™ Supplier No. MA528662
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Qa-2 Monoclonal Antibody for Flow

Qa-2 is a GPI-linked MHC class I beta surface molecule. Qa-2 is expressed by mature T and B cells, and its expression level varies between laboratory mouse strains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Qa-2
Applications Flow Cytometry
Classification Monoclonal
Clone 695H1-9-9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation BBS with proprietary stabilizer and no preservative; pH 8.2
Gene Qa2
Gene Accession No. 0
Gene Alias H2Q7; H2-Qa2; Ped; Qa lymphocyte antigen 2 region; Qa2; Qa-2; Qa-2 antigen
Gene Symbols Qa2
Host Species Mouse
Immunogen C3H.SW mouse skin graft and splenocytes.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 111458
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.