Learn More
Microbiologics™ QA/QC Microorganism Preparations in KWIK-STIK™ Plus
Shop All Microbiologics Products

Description
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
‡
Biotype G
Specifications
Specifications
| ATCC® | 14028 |
| Biosafety Level | 2 |
| Format | KWIK-STIK™ Plus |
| Product Type | KWIK-STIK |
| Packaging Type | Canister |
| Quantity | 5 units of a single strain |
| Species | Salmonella typhimurium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.