Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QA1b Antibody (6A8.6F10.1A6), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP226650
Description
QA1b Monoclonal specifically detects QA1b in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
QA1b | |
Monoclonal | |
Biotin | |
NP_001129540 | |
QA1b | |
Animals were immunized with a synthetic peptide from the alpha-2 domain of Qa-1b. Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells. Spleen cells were fused with P3X63Ag8.653 myeloma cells.The antibody reacts with the Qa-1b Class Ib molecule. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
6A8.6F10.1A6 | |
Flow Cytometry 1ul/1 million cells | |
H2-Q1, H2-T23 | |
Mouse | |
Protein G purified | |
RUO | |
15040 | |
Store at 4C. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction