Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QA1b Antibody (6A8.6F10.1A6) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen QA1b
Clone 6A8.6F10.1A6
Concentration 1.0 mg/ml
Dilution Western Blot 1 - 2 ug/ml, Flow Cytometry 1:10 - 1:1000
Applications Western Blot, Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP226649SS
SDP
View Documents
Novus Biologicals
NBP226649SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP226649
SDP
View Documents
Novus Biologicals
NBP226649
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

QA1b Monoclonal specifically detects QA1b in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Neutralization.
Specifications

Specifications

QA1b
1.0 mg/ml
Western Blot, Flow Cytometry
Unconjugated
Mouse
Human, Mouse
H2-Q1, H2-T23
QA1b
IgG1 κ
Protein G purified
6A8.6F10.1A6
Western Blot 1 - 2 ug/ml, Flow Cytometry 1:10 - 1:1000
Monoclonal
Purified
RUO
NP_001129540
15040
Animals were immunized with a synthetic peptide from the alpha-2 domain of Qa-1b. Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells. Spleen cells were fused with P3X63Ag8.653 myeloma cells. The antibody reacts with the Qa-1b Class Ib molecule.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.