Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QA1b Antibody (6A8.6F10.1A6), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP226649AF350
Description
QA1b Monoclonal specifically detects QA1b in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
QA1b | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
QA1b | |
Animals were immunized with a synthetic peptide from the alpha-2 domain of Qa-1b. Qa-1 is a Class Ib molecule recognized by cytotoxic T cells and natural killer cells. Spleen cells were fused with P3X63Ag8.653 myeloma cells. The antibody reacts with the Qa-1b Class Ib molecule. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 κ |
Western Blot, Flow Cytometry | |
6A8.6F10.1A6 | |
Western Blot, Flow Cytometry | |
H2-Q1, H2-T23 | |
Mouse | |
Protein G purified | |
Signal Transduction | |
15040.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction