Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QBC Diagnostics F.A.S.T. Prepackaged AFB Kits

Catalog No. p-4374923 Shop All Drucker Diagnostics Products
Click to view available options
Type:
F.A.S.T AFB Smear Stain Kit
F.A.S.T. AFB Smear Kit with Digestion Solution

Catalog No. 23-900-144

Supplier: QBC Diagnostics 427409

Includes

QBC F.A.S.T AFB Smear Kit

  • F.A.S.T Auramine O Stain Kit (120mL)
  • Quality control slides (5)
  • F.A.S.T SureFocus Miscroscope Slides (432)
  • F.A.S.T Sputum Cups (400)
  • F.A.S.T Applicator Sticks (400)
QBC F.A.S.T AFB Smear Kit with Digestion Solution
  • F.A.S.T Auramine O Stain Kit (120mL)
  • Quality control slides (5)
  • F.A.S.T SureFocus Miscroscope Slides (432)
  • Bottles of F.A.S.T Sputum Digestion Solution (27)

Complete AFB staining systems

  • Three minute staining
  • Supplies for 400 tests

Specifications

Type F.A.S.T AFB Smear Stain Kit
Quantity 400 Tests
Includes 120mL Auramine O stain kit, 5 quality control slides, 432 SureFocus miscroscope slides, 400 sputum cups, 400 applicator sticks
For Use With (Application) Detect positive AFB (tuberculosis) sputum samples
Storage Requirements 15° to 25°C
Stain Type Auramine O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.