Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ QC Sets and Panels: Helix Elite™
Shop All Microbiologics Products

Click to view available options
Quantity:
6 Pellets
Description
- Shipping restriction: Not available for shipment to Canada or Europe
- Test Method: Microbial Identification, Molecular Testing
- Permits and Restrictions: Export Restrictions May Applyon.
Respiratory (21 Targets) Control Panel contains six positive controls in a pre-pooled inactivated pellet format and six negative control pellets (12 vials/pellets total). The negative control pellets are blank and do not contain microorganisms.
Positive Control (six vials/pellets):
- Adenovirus Type 6
- Bordetella parapertussis
- Bordetella pertussis
- Chlamydia pneumoniae
- Coronavirus 229E
- Recombinant Coronavirus HKU1
- Recombinant Coronavirus NL63
- Recombinant Coronavirus OC43 Strain 1 and Recombinant Coronavirus OC43 Strain 2
- Recombinant Human Metapneumovirus
- Human Rhinovirus
- Influenza A
- Influenza A subtype H1
- Influenza A subtype H1-2009
- Influenza A subtype H3
- Influenza B
- Mycoplasma pneumoniae
- Parainfluenza Virus 1
- Parainfluenza Virus 2
- Parainfluenza Virus 3
- Recombinant Parainfluenza Virus 4a
- Respiratory Syncytial Virus
Negative Control (six vials/pellets): Blank pellets
Specifications
Specifications
| Biosafety Level | 1 |
| Content And Storage | 2 to 25°C |
| Format | Helix Elite |
| Manufacturing Origin | United States |
| Product Type | Helix Elite Control Reference Panels and Standards |
| For Use With (Application) | Microbial Identification, Molecular Testing |
| Packaging Type | Plastic Box |
| Quantity | 6 Pellets |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction