Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QDPR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563632 Supplier Invitrogen™ Supplier No. PA563632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLAGKNSGMP PGAAAIAVLP VTLDTPMNRK SMPEADFSSW TPLEFLVETF HDWITGKNRP SSGSLIQVVT TEGRTEL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

This gene encodes the enzyme dihydropteridine reductase, which catalyzes the NADH-mediated reduction of quinonoid dihydrobiopterin. This enzyme is an essential component of the pterin-dependent aromatic amino acid hydroxylating systems. Mutations in this gene resulting in QDPR deficiency include aberrant splicing, amino acid substitutions, insertions, or premature terminations. Dihydropteridine reductase deficiency presents as atypical phenylketonuria due to insufficient production of biopterin, a cofactor for phenylalanine hydroxylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QDPR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene QDPR
Gene Accession No. P09417
Gene Alias 2610008L04Rik; 6,7-dihydropteridine reductase; D5Ertd371e; Dhpr; dihydropteridine reductase; HDHPR; HPR; PKU2; QDPR; Quinoid dihydropteridine reductase; SDR33C1; Short chain dehydrogenase/reductase family 33C member 1; short chain dehydrogenase/reductase family 33C, member 1; testis secretory sperm-binding protein Li 236P
Gene Symbols QDPR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human QDPR. Recombinant protein control fragment (Product #RP-101362).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5860
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.