Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ QDPR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564233
Description
Immunogen sequence: WVASVDVVEN EEASASIIVK MTDSFTEQAD QVTAEVGKLL GEEKVDAILC VAGGWAGGNA KSKSLFKNCD LMWKQSIWTS TISSHLATKH Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 96%.
This gene encodes the enzyme dihydropteridine reductase, which catalyzes the NADH-mediated reduction of quinonoid dihydrobiopterin. This enzyme is an essential component of the pterin-dependent aromatic amino acid hydroxylating systems. Mutations in this gene resulting in QDPR deficiency include aberrant splicing, amino acid substitutions, insertions, or premature terminations. Dihydropteridine reductase deficiency presents as atypical phenylketonuria due to insufficient production of biopterin, a cofactor for phenylalanine hydroxylase.
Specifications
QDPR | |
Polyclonal | |
Unconjugated | |
QDPR | |
2610008L04Rik; 6,7-dihydropteridine reductase; D5Ertd371e; Dhpr; dihydropteridine reductase; HDHPR; HPR; PKU2; QDPR; Quinoid dihydropteridine reductase; SDR33C1; Short chain dehydrogenase/reductase family 33C member 1; short chain dehydrogenase/reductase family 33C, member 1; testis secretory sperm-binding protein Li 236P | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5860 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09417 | |
QDPR | |
Recombinant protein corresponding to Human QDPR. Recombinant protein control fragment (Product #RP-103971). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction