Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QDPR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110250 Supplier Invitrogen™ Supplier No. PA5110250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAAAAGEA RRVLVYGGRG ALGSRCVQAF RARNWWVASV DVVENEEASA SIIVKMTDSF TEQADQVTAE VGKLLGEEKV DAILCVAGGW AGGNAKSKSL FKNCDLMWKQ SIWTSTISSH LATKHLKEGG LLTLAGAKAA LDGTPGMIGY GMAKGAVHQL CQSLAGKNSG MPPGAAAIAV LPVTLDTPMN RKSMPEADFS SWTPLEFLVE TFHDWITGKN RPSSGSLIQV VTTEGRTELT PAYF.

This gene encodes the enzyme dihydropteridine reductase, which catalyzes the NADH-mediated reduction of quinonoid dihydrobiopterin. This enzyme is an essential component of the pterin-dependent aromatic amino acid hydroxylating systems. Mutations in this gene resulting in QDPR deficiency include aberrant splicing, amino acid substitutions, insertions, or premature terminations. Dihydropteridine reductase deficiency presents as atypical phenylketonuria due to insufficient production of biopterin, a cofactor for phenylalanine hydroxylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QDPR
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene QDPR
Gene Accession No. P09417, P11348, Q8BVI4
Gene Alias 2610008L04Rik; 6,7-dihydropteridine reductase; D5Ertd371e; Dhpr; dihydropteridine reductase; HDHPR; HPR; PKU2; QDPR; Quinoid dihydropteridine reductase; SDR33C1; Short chain dehydrogenase/reductase family 33C member 1; short chain dehydrogenase/reductase family 33C, member 1; testis secretory sperm-binding protein Li 236P
Gene Symbols QDPR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-244 of human QDPR (NP_000311.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110391, 5860, 64192
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.