Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Qiagen™ Robotic Tips

Catalog No. 14222387
Click to view available options
Volume (Metric):
1000 μL
Sterility:
Sterile
Color:
Black

Compatible with Qiagen liquid handling systems and Qiagen BioRobot Series

Available in volumes of 250, 300, 1000 and 1100 μL.

  • Clear and conductive black tips available
  • Choice of Presterilized, Nonsterilized, Filtered, Non-Filtered and Conductive tips

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Black
Certifications/Compliance Non-ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, liquid level sensing, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 16 Racks/Cs.
For Use With (Equipment) Qiagen BioRobot
Product Line Qiagen
Material Polyethylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.