Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIK1/SIK2/SIK3 Polyclonal Antibody
SIK2/SIK3 Polyclonal Antibody
Supplier: Thermo Scientific PA5106254
Description
SIK2/SIK3 Polyclonal Antibody for Western Blot

Specifications
SIK1/SIK2/SIK3 | |
Polyclonal | |
Unconjugated | |
SIK1 | |
QIK; QSK | |
Rabbit | |
Affinity Chromatography | |
RUO | |
150094, 17691, 23235, 23387, 235344, 315649, 59329, 684112, 70661 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P57059, Q60670, Q6P4S6, Q8CFH6, Q9H0K1, Q9R1U5, Q9Y2K2 | |
SIK1, SIK2, SIK3 | |
A synthesized peptide derived from human SIK1(Accession P57059), corresponding to amino acid residues K164-L214. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction