Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QIL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560074
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560074 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560074 Supplier Invitrogen™ Supplier No. PA560074
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VYLVYDQELL GPSDKSQAAL QKAGEVVPPA MYQFSQYVCQ QTGLQIPQLP APPKIYFPIR DSWNAGIMTV MSALSVAPSK AREYSKEGWE YVK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 23%.

Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Constituent of mature MICOS complex, it is required for the formation of cristae junction (CJ) and maintenance of cristae morphology. Required for the incorporation of MICOS10/MIC10 into the MICOS complex. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen QIL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MICOS13
Gene Accession No. Q5XKP0
Gene Alias 2410015M20Rik; C19orf70; chromosome 19 open reading frame 70; MIC13; MICOS complex subunit MIC13; Micos13; P117; Protein P117; protein QIL1; QIL1; RIKEN cDNA 2410015M20 gene; sr104
Gene Symbols MICOS13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human QIL1. Recombinant protein control fragment (Product #RP-99183).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.