Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QKI Monoclonal Antibody (GT1589)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527804

Catalog No. PIMA527804


Only null left
Add to Cart

Description

Description

Positive Control: 293T Predicted Reactivity: Xenopus laevis (93%), Dog (100%), Cat (100%), Pig (100%), Chicken (98%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes the sacsin protein, which includes a UbL domain at the N-terminus, a DnaJ domain, and a HEPN domain at the C-terminus. The gene is highly expressed in the central nervous system, also found in skin, skeletal muscles and at low levels in the pancreas. This gene includes a very large exon spanning more than 12.8 kb. Mutations in this gene result in autosomal recessive spastic ataxia of Charlevoix-Saguenay (ARSACS), a neurodegenerative disorder characterized by early-onset cerebellar ataxia with spasticity and peripheral neuropathy. Sacsin has been found to protect against mutant ataxin-1. A pseudogene associated with this gene is located on chromosome 11. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

QKI
Monoclonal
1 mg/mL
PBS with no preservative
Q91XU1, Q96PU8, Q9QYS9
Qk, QKI
Recombinant protein encompassing a sequence within the center region of human QKI. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
GT1589
Unconjugated
QKI
1110003F05Rik; 1500005P18; DKFZp586I0923; HKQ; homolog of mouse quaking QKI (KH domain RNA binding protein); Hqk; hqkI; l(17)-1Wis; l17Wis1; lethal, Chr 17, U Wisconsin 1; MqkI; Protein quaking; protein quaking-like; Qk; QK1; QK3; Qka1; Qki; QKI, KH domain containing RNA binding; QKI, KH domain containing, RNA binding; QKI-5 protein; quaking; quaking homolog, KH domain RNA binding; RNA Binding (QKI); RNA binding protein HQK; rqkI
Mouse
Protein A
RUO
100910212, 19317, 499022, 9444
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.