Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QKI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598704 Supplier Invitrogen™ Supplier No. PA598704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

QKI Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes the sacsin protein, which includes a UbL domain at the N-terminus, a DnaJ domain, and a HEPN domain at the C-terminus. The gene is highly expressed in the central nervous system, also found in skin, skeletal muscles and at low levels in the pancreas. This gene includes a very large exon spanning more than 12.8 kb. Mutations in this gene result in autosomal recessive spastic ataxia of Charlevoix-Saguenay (ARSACS), a neurodegenerative disorder characterized by early-onset cerebellar ataxia with spasticity and peripheral neuropathy. Sacsin has been found to protect against mutant ataxin-1. A pseudogene associated with this gene is located on chromosome 11. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QKI
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene QKI
Gene Accession No. Q96PU8
Gene Alias 1110003F05Rik; 1500005P18; DKFZp586I0923; HKQ; homolog of mouse quaking QKI (KH domain RNA binding protein); Hqk; hqkI; l(17)-1Wis; l17Wis1; lethal, Chr 17, U Wisconsin 1; MqkI; Protein quaking; protein quaking-like; Qk; QK1; QK3; Qka1; Qki; QKI, KH domain containing RNA binding; QKI, KH domain containing, RNA binding; QKI-5 protein; quaking; quaking homolog, KH domain RNA binding; RNA Binding (QKI); RNA binding protein HQK; rqkI
Gene Symbols QKI
Host Species Rabbit
Immunogen Recombinant Human Protein quaking protein (1-341AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9444
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.