Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QKI/Quaking Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300239AF647
Description
QKI/Quaking Polyclonal antibody specifically detects QKI/Quaking in Human samples. It is validated for Western BlotSpecifications
QKI/Quaking | |
Polyclonal | |
Western Blot | |
DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
A synthetic peptide mapping to a region between residues 50 and 100 of isoform 5 of human quaking homolog using the numbering given in entry AAH19917.1 (GeneID 9444). | |
0.1 mL | |
Neuroscience | |
9444 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction