Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QKI/Quaking Antibody (GT2612), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313576
This item is not returnable.
View return policy
Description
QKI/Quaking Monoclonal antibody specifically detects QKI/Quaking in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| QKI/Quaking | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| GT2612 | |
| Western Blot, Immunohistochemistry-Paraffin 5ug/ml | |
| DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
| Recombinant protein encompassing a sequence within the center region of human QKI/Quaking. The exact sequence is proprietary. | |
| 100 μg | |
| Neuroscience | |
| 9444 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction