Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QKI/Quaking Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB300239
Description
QKI/Quaking Polyclonal specifically detects QKI/Quaking in Human samples. It is validated for Western Blot.Specifications
QKI/Quaking | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
Rabbit | |
38 kDa | |
100 μL | |
Neuroscience | |
9444 | |
Human | |
IgG |
Western Blot | |
1.0 mg/mL | |
Western Blot 1:1000-1:5000 | |
Q96PU8 | |
QKI | |
A synthetic peptide mapping to a region between residues 50 and 100 of isoform 5 of human quaking homolog using the numbering given in entry AAH19917.1 (GeneID 9444). | |
Affinity Purified | |
RUO | |
Primary | |
The epitope recognized is common to isoforms 1, 2, 3, 4, 5, 6, 7 and 7B according to TrEMBL entries Q9P0Y1, Q9P0Y0, Q9P0X6, Q9P0X8, Q96PU8, Q96PU7, Q969L9 and Q96PU6, respectively. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction