Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QKI/Quaking Antibody (PCRP-QKI-2F10), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314072AF647
This item is not returnable.
View return policy
Description
QKI/Quaking Monoclonal antibody specifically detects QKI/Quaking in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| QKI/Quaking | |
| Monoclonal | |
| Alexa Fluor 647 | |
| DKFZp586I0923, HKQ, homolog of mouse quaking QKI (KH domain RNA binding protein), Hqk, hqkI, protein quaking, QK, QK1, QK3, quaking homolog, KH domain RNA binding (mouse), RNA binding protein HQK | |
| Recombinant full-length human QKI/Quaking protein (Uniprot: Q96PU8) | |
| 0.1 mL | |
| Neuroscience | |
| 9444 | |
| Store at 4°C in the dark. | |
| IgG2c |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-QKI-2F10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction