Learn More
Thermo Scientific™ QMS™ Tacrolimus Immunoassays and Calibrators
GSA_VA
Catalog No. p-6161843
Change view
Click to view available options
Type:
Tacrolimus Calibrators
Tacrolimus Immunoassay
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-278-445 Tacrolimus Immunoassay
23-278-444 Tacrolimus Calibrators
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-445 Supplier Thermo Scientific™ Supplier No. 10015556
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Achieve excellent low-end precision and sensitivity with the Thermo Scientific™ QMS™ Tacrolimus Immunoassay.
Achieve high sensitivity and excellent low-end precision when monitoring kidney, liver and heart transplant patients with the Thermo Scientific™ QMS™ Tacrolimus Immunoassay. With a reportable range down to 1ng/mL, our Tacrolimus Immunoassay is the perfect aid for monitoring patients on multiple drug therapies.The reagents are liquid and ready-to-use with applications available on a wide variety of other laboratory instrumentation. List of applications available upon request.
Key Features:
  • ISD Assays utilize a recombinant DNA technology to produce a unique homogeneous enzyme immunoassay system
  • QMS ISD and TDM Assays use coated microparticle beads
  • Technology is based on competitive inhibition principles and photometrically measures rate of absorbance change
Accurate:
  • Excellent low-end sensitivity
  • Correlation with gold standard methods
  • Precise throughput standard curve assay range
  • Lot-to-lot consistency
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description QMS Tacrolimus Immunoassay for Indiko
Detectable Analytes Tacrolimus
Sample Type Whole Blood
Type Tacrolimus Immunoassay
Quantity 18 mL R1, 12 mL R2, 50 mL extraction