Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Controls
Click to view available options
Description:
Lamotrigine Controls
Topiramate Controls
Zonisamide Controls
Description
Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Zonisamide Controls |
| Concentration | 8.0, 25.0, 50.0 μg/mL |
| Detectable Analytes | Zonisamide |
| Sample Type | Plasma, Serum |
| Type | Control |
| Level | Level 3 |
| Quantity | 3 x 2.5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction