Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays

Catalog No. 23279101
Change view
Click to view available options
Quantity:
1 x 14 mL, 1 x 12 mL
1 x 19 mL, 1 x 14 mL
1 x 19 mL, 1 x 7 mL
1 x 19 mL, 1 x 8 mL
1 x 22 mL, 1 x 16 mL
1 x 22 mL, 1 x 9 mL
2 x 19 mL
2 x 19 mL, 2 x 7 mL
2 x 19 mL, 2 x 8 mL
2 x 22 mL
2 x 22 mL, 2 x 8 mL
Detectable Analytes:
Amikacin
Gentamicin
Lamotrigine
Quinidine
Tobramycin
Topiramate
Vancomycin
Zonisamide
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Detectable Analytes
23-279-101 2 x 19 mL, 2 x 7 mL Quinidine
23-278-242 2 x 19 mL, 2 x 7 mL Amikacin
21-200-419 1 x 22 mL, 1 x 9 mL Gentamicin
23-279-102 2 x 19 mL Lamotrigine
23-279-103 1 x 22 mL, 1 x 16 mL Topiramate
21-200-440 2 x 22 mL Vancomycin
23-279-100 2 x 22 mL, 2 x 8 mL Zonisamide
23-278-451 1 x 19 mL, 1 x 7 mL Amikacin
23-278-449 1 x 19 mL, 1 x 8 mL Gentamicin
23-278-450 1 x 14 mL, 1 x 12 mL Tobramycin
23-278-453 1 x 19 mL, 1 x 14 mL Topiramate
23-278-454 2 x 19 mL Vancomycin
23-278-455 2 x 19 mL, 2 x 8 mL Zonisamide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Catalog No. 23-279-101 Supplier Thermo Scientific™ Supplier No. 0373936
Add to Cart
Add to Cart
This item is not returnable. View return policy

Utilize the precision, consistency and optimized performance of Thermo Scientific™ QMS™ TDM Assays, ready-to-use, liquid immunoassays.

Accurately measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays. These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.

Features:
  • Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
  • Liquid stable and ready to use
  • Applicable to general chemistry analyzers
  • Validated parameters assure lab-to-lab reproducibility
  • Superb reagent stability
  • Excellent correlation to reference methods
  • Optimal sensitivity, precision and dynamic range
  • Low cross-reactivity with metabolites and commonly co-administered drugs
  • Low interference from endogenous substances
  • Based on the competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles.

 

Includes:

Two reagent system, 2 x 19, 2 x 7mL

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Quinidine Assay
Detectable Analytes Quinidine
Sample Type Plasma, Serum
Type Reagent
Quantity 2 x 19 mL, 2 x 7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.