Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays

Description
Accurately measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays. These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.
- Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
- Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
- Liquid stable and ready-to-use
- Applicable to general chemistry analyzers
- Validated parameters assure lab-to-lab reproducibility
- Superb reagent stability
- Excellent correlation to reference methods
- Optimal sensitivity, precision and dynamic range
- Low cross-reactivity with metabolites and commonly co-administered drugs
- Low interference from endogenous substances
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Lamotrigine Assay |
| Detectable Analytes | Lamotrigine |
| Sample Type | Plasma, Serum |
| Type | Reagent |
| Quantity | 2 x 19 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.