Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ QNZ 46
GREENER_CHOICESDP

Catalog No. 480150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-015-0 50 mg
48-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-015-0 Supplier R&D Systems™ Supplier No. 4801/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. NR2C/NR2D-selective NMDA receptor non-competitive antagonist

GluN2C/GluN2D-selective NMDA receptor non-competitive antagonist (IC50 values are 3, 6, 229, and >300, >300 μM for GluN2D, GluN2C, GluN2A, GluN2B, and GluR1, respectively). Requires binding of glutamate to the GluN2 subunit.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 1237744-13-6
Molecular Formula C24H17N3O6

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 443.41
Purity >98%
Molecular Weight (g/mol) 443.415
Chemical Name or Material QNZ 46
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.