Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Polypropylene SturdeeSeal PE Foam Lined Caps

Catalog No. 02991068
Click to view available options
For Use With (Equipment):
Bottle
Wide mouth bottle
Neck Finish:
120-400
20-410
24-410
28-410
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
144 Cs.
1500 Cs.
1650 Cs.
2000 Cs.
210 Cs.
2500 Cs.
2700 Cs.
288 Cs.
3000 Cs.
3750 Cs.
4300 Cs.
5000 Cs.
576 Cs.
608 Cs.
7200 Cs.
760 Cs.
Packaging:
Bagged
Bulk

Features excellent sealing properties

  • Can be used when good impact strength and pliability are required
  • Resistant to many chemicals including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
  • Available in ribbed style
  • Available in bulk or bagged packing

Specifications

Color White
Material Polypropylene
No. per Case 608 Cs.
Packaging Bulk
Neck Finish 89-400
For Use With (Equipment) Wide mouth bottle
Closure Liner Polyethylene Foam
Closure Type Ribbed
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.