Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Alpha Caps for F-Style Cans
Click to view available options
Liner Material:
Pulp/Aluminum Foil
Solvseal
Description
- Impact resistant and shows wide range of temperature tolerances
- Available with pulp/alum foil or Solvseal liner
- Solvseal liners are ideal for vegetable and mineral oils, weak acids and alkalis, alcohols and organic solvents
- Aluminum foil lined caps are used for food applications and can be used for non-acid, non-alkaline products
Specifications
Specifications
| Color | Silver |
| Material | Metal |
| No. per Case | 2700 |
| Liner Material | Pulp/Aluminum Foil |
| For Use With (Equipment) | Vegetable and mineral oil |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction